1-(Methoxymethyl)-3-(trifluoromethyl)pyrazole-4-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1-(Methoxymethyl)-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid

A337191-250MG A337191-100MG A337191-1G
77890-674EA 190 USD
77890-674 77890-672 77890-673
1-(Methoxymethyl)-3-(trifluoromethyl)pyrazole-4-boronic acid ≥98%
1-(Methoxymethyl)-3-(trifluoromethyl)pyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BF₃N₂O₃
MDL Number: MFCD15143477
CAS Number: 1218790-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR