Oxan-3-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: Tetrahydro-2H-pyran-3-amine

A330059-100MG A330059-1G A330059-250MG
77890-657EA 62 USD
77890-657 77890-658 77890-659
Oxan-3-amine ≥98%
Oxan-3-amine

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.147 g/mol
MDL Number: MFCD07371531
CAS Number: 120811-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR