Resminostat (hydrochloride) ≥98%

Supplier: Ambeed
Warning

A336907-10MG A336907-1MG A336907-5MG
77616-486EA 105.06 USD
77616-486 77616-488 77616-490
Resminostat (hydrochloride) ≥98%
Resminostat (hydrochloride)

Precaución: for Research Use Only

Formula: C₁₆H₂₀ClN₃O₄S
MDL Number: MFCD26142920
CAS Number: 1187075-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR