Cyclopentadienylmanganese tricarbonyl ≥98%

Supplier: Ambeed
Danger

Synonyms: Cymantrene, Cyclopentadienylmanganese(I) tricarbonyl

A329851-1G A329851-250MG
77601-484EA 80.98 USD
77601-484 77626-630
Cyclopentadienylmanganese tricarbonyl ≥98%
Cyclopentadienylmanganese tricarbonyl

Precaución: for Research Use Only

Formula: C₈H₅MnO₃
MW: 204.06 g/mol
MDL Number: MFCD00001440
CAS Number: 12079-65-1
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR