1-(1-Naphthylmethyl)piperazine ≥98%

Supplier: Ambeed
Danger

Synonyms: 1-(Naphthalen-1-ylmethyl)piperazine

A336396-250MG A336396-5G
77872-489EA 14 USD
77872-489 77872-490
1-(1-Naphthylmethyl)piperazine ≥98%
1-(1-Naphthylmethyl)piperazine

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂
MW: 226.323 g/mol
MDL Number: MFCD01314185
CAS Number: 40675-81-8
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR