meso-Hydrobenzoin ≥98%

Supplier: Ambeed
Warning

A329380-5G A329380-1G
77593-948EA 58.43 USD
77593-948 77879-696
meso-Hydrobenzoin ≥98%
meso-Hydrobenzoin

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
MDL Number: MFCD00064253
CAS Number: 579-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR