Methyl N-acetyl-α-D-glucosaminide ≥95%

Supplier: Ambeed
Warning

A329314-1G A329314-250MG A329314-5G
77626-626EA 135.96 USD
77626-626 77626-628 77878-035
Methyl N-acetyl-α-D-glucosaminide ≥95%
Methyl N-acetyl-α-D-glucosaminide

Precaución: for Research Use Only

Formula: C₉H₁₇NO₆
MDL Number: MFCD00056076
CAS Number: 6082-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR