CFI-400945 ≥98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-2-(3-((E)-4-(((2R,6S)-2,6-Dimethylmorpholino)methyl)styryl)-1H-indazol-6-yl)-5'-methoxyspiro[cyclopropane-1,3'-indolin]-2'-one, (1R,2S)-2-(3-((E)-4-(((2S,6R)-2,6-dimethylmorpholino)methyl)styryl)-1H-indazol-6-yl)-5'-methoxyspiro[cyclopropane-1,3'-indolin]-2'-one

A335666-5MG A335666-100MG A335666-50MG
77685-959EA 81 USD
77685-959 77892-693 77892-694
CFI-400945 ≥98%
CFI-400945

Precaución: for Research Use Only

Formula: C₃₃H₃₄N₄O₃
MDL Number: MFCD28386170
CAS Number: 1338806-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR