2-(3-(3-(5-Hydroxypyridin-2-yl)-1,2,4-oxadiazol-5-yl)-2,2-dimethylpropanamido)cyclohex-1-ene-1-carboxylic acid ≥99%

Supplier: Ambeed
Warning

A335426-1MG A335426-10MG A335426-5MG
77616-478EA 61.8 USD
77616-478 77685-957 77685-958
2-(3-(3-(5-Hydroxypyridin-2-yl)-1,2,4-oxadiazol-5-yl)-2,2-dimethylpropanamido)cyclohex-1-ene-1-carboxylic acid ≥99%
2-(3-(3-(5-Hydroxypyridin-2-yl)-1,2,4-oxadiazol-5-yl)-2,2-dimethylpropanamido)cyclohex-1-ene-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₂₂N₄O₅
MDL Number: MFCD20528236
CAS Number: 917910-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR