4-Phenyl-1H-pyrrole-3-carboxylic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A335070-100MG A335070-1G A335070-250MG
77684-002EA 14 USD
77684-002 77890-669 77890-670
4-Phenyl-1H-pyrrole-3-carboxylic acid ≥98%, Ambeed.Inc
4-Phenyl-1H-pyrrole-3-carboxylic acid

Precaución: For research use only.

Formula: C₁₁H₉NO₂
MDL Number: MFCD06411253
CAS Number: 132040-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR