Benzyl 4-(4-((tert-butoxycarbonyl)amino)cyclohexyl)piperazine-1-carboxylate ≥95%
Supplier: Ambeed
|
Warning
|
A326613-250MG
A326613-1G
A326613-100MG
77612-882EA
79.31
USD
77612-882
77683-974
77683-973
Benzyl 4-(4-((tert-butoxycarbonyl)amino)cyclohexyl)piperazine-1-carboxylate ≥95%
Benzyl 4-(4-((tert-butoxycarbonyl)amino)cyclohexyl)piperazine-1-carboxylate
Precaución: For research use only.
Formula:
C₂₃H₃₅N₃O₄ MW: 417.54 g/mol |
MDL Number:
MFCD23115353 CAS Number: 1248730-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...