Fleroxacin 98+%

Supplier: Ambeed
Warning

Synonyms: Megalone, 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid, 6,8-Difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

A328492-5G A328492-100G A328492-25G
77601-480EA 56.38 USD
77601-480 77668-480 77668-482
Fleroxacin 98+%
Fleroxacin

Precaución: for Research Use Only

Formula: C₁₇H₁₈F₃N₃O₃
MW: 369.34 g/mol
MDL Number: MFCD00864880
CAS Number: 79660-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR