(1R,2R)-1-(4-Nitrophenyl)-2-tetradecanamidopropane-1,3-diyl bis(2-(dimethylamino)acetate) dihydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: LCL521 2HCl

A328422-10MG A328422-1MG A328422-5MG
77616-442EA 103 USD
77616-442 77616-444 77616-446
(1R,2R)-1-(4-Nitrophenyl)-2-tetradecanamidopropane-1,3-diyl bis(2-(dimethylamino)acetate) dihydrochloride ≥98%
(1R,2R)-1-(4-Nitrophenyl)-2-tetradecanamidopropane-1,3-diyl bis(2-(dimethylamino)acetate) dihydrochloride

Precaución: for Research Use Only

Formula: C₃₁H₅₄Cl₂N₄O₇
MDL Number: MFCD31692338
CAS Number: 1226759-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR