3-Trifluoromethylpyrrolidine hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A328261-1G
A328261-250MG
A328261-5G
77883-892EA
124
USD
77883-892
77883-893
77883-894
3-Trifluoromethylpyrrolidine hydrochloride ≥98%
3-Trifluoromethylpyrrolidine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₉ClF₃N |
MDL Number:
MFCD03452798 CAS Number: 1189485-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...