(1-Methyl-1H-pyrrol-2-yl)methanamine ≥95%

Supplier: Ambeed
Warning

Synonyms: [(1-Methyl-1H-pyrrol-2-yl)methyl]amine, 2-(Aminomethyl)-1-methyl-1H-pyrrole

A327644-100MG A327644-1G
77685-938EA 61 USD
77685-938 77685-939
(1-Methyl-1H-pyrrol-2-yl)methanamine ≥95%
(1-Methyl-1H-pyrrol-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₁₀N₂
MW: 110.15 g/mol
MDL Number: MFCD02677708
CAS Number: 69807-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR