Trimethoprim ≥98%

Supplier: Ambeed
Warning

A327622-100G A327622-25G A327622-500G A327622-5G A327622-1000G
77114-832EA 45.33 USD
77114-832 77114-834 77114-836 77114-838 77685-937
Trimethoprim ≥98%
Trimethoprim

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₄O₃
MDL Number: MFCD00036761
CAS Number: 738-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR