(R)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride ≥98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A327611-5G
A327611-1G
77683-976EA
47
USD
77683-976
77892-676
(R)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride ≥98%, Ambeed.Inc
(R)-2-Methylpyrrolidine-2-carboxylic acid hydrochloride
Precaución: For research use only.
Formula:
C₆H₁₂ClNO₂ MW: 165.62 g/mol |
MDL Number:
MFCD20487714 CAS Number: 123053-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...