5-Fluoro-3-methylsalicylaldehyde ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

A327458-5G A327458-1G
77683-975EA 108 USD
77683-975 77890-650
5-Fluoro-3-methylsalicylaldehyde ≥95%, Ambeed.Inc
5-Fluoro-3-methylsalicylaldehyde

Precaución: For research use only.

Formula: C₈H₇FO₂
MDL Number: MFCD04039244
CAS Number: 704884-74-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR