(5-Amino-2-nitrophenyl)methanol ≥95%

Supplier: Ambeed
Warning

A327440-1G A327440-250MG
77883-890EA 200 USD
77883-890 77883-891
(5-Amino-2-nitrophenyl)methanol ≥95%
(5-Amino-2-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD00036068
CAS Number: 77376-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR