Nonadecanoic acid methyl ester ≥98%

Supplier: Ambeed
Warning

A326895-1G A326895-250MG A326895-25G A326895-5G A326895-100MG
77668-470EA 22 USD
77668-470 77668-472 77668-474 77668-476 77879-685
Nonadecanoic acid methyl ester ≥98%
Nonadecanoic acid methyl ester

Precaución: for Research Use Only

Formula: C₂₀H₄₀O₂
MDL Number: MFCD00009011
CAS Number: 1731-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR