4-Carboxy-2-methoxyphenylboronic acid ≥95%

Supplier: Ambeed
Warning

A326719-100MG A326719-10G A326719-1G A326719-250MG A326719-5G
77883-885EA 23 USD
77883-885 77883-886 77883-887 77883-888 77883-889
4-Carboxy-2-methoxyphenylboronic acid ≥95%
4-Carboxy-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MDL Number: MFCD10696665
CAS Number: 741699-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR