2-Bromo-6-fluoro-3-methoxybenzeneboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-6-fluoro-3-methoxyphenyl)boronic acid

A318531-1G A318531-25G A318531-5G
77881-893EA 8 USD
77881-893 77881-894 77881-895
2-Bromo-6-fluoro-3-methoxybenzeneboronic acid ≥98%
2-Bromo-6-fluoro-3-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₃
MDL Number: MFCD08276828
CAS Number: 957062-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR