2-Chloro-1,3-bis(methoxycarbonyl)guanidine ≥95%

Supplier: Ambeed
Danger

A318241-5G A318241-1G
77593-888EA 111.73 USD
77593-888 77683-946
2-Chloro-1,3-bis(methoxycarbonyl)guanidine ≥95%
2-Chloro-1,3-bis(methoxycarbonyl)guanidine

Precaución: For research use only.

Formula: C₅H₈ClN₃O₄
MW: 209.58 g/mol
MDL Number: MFCD28016343
CAS Number: 1596379-00-8
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR