Lycorine HCl ≥99%

Supplier: Ambeed
Danger

Synonyms: (1S,2S,3a1S,12bS)-2,3a1,4,5,7,12b-Hexahydro-1H-[1,3]dioxolo[4,5-j]pyrrolo[3,2,1-de]phenanthridine-1,2-diol hydrochloride, Lycorine hydrochloride

A318231-1G A318231-5G A318231-100MG A318231-1MG A318231-25G A318231-50MG
77612-844EA 81.37 USD
77612-844 77612-846 77685-902 77685-903 77685-904 77685-905
Lycorine HCl ≥99%
Lycorine HCl

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₄·HCl
MW: 323.8 g/mol
MDL Number: MFCD00243111
CAS Number: 2188-68-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR