2,5-difluoro-3-nitrotoluene ≥98%

Supplier: Ambeed
Danger

Synonyms: 2,5-Difluoro-1-methyl-3-nitrobenzene

A318038-5G A318038-1G A318038-25G
77672-630EA 74 USD
77672-630 77881-891 77881-892
2,5-difluoro-3-nitrotoluene ≥98%
2,5-difluoro-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MDL Number: MFCD11519028
CAS Number: 1093758-82-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR