Phenylselenol ≥98%

Supplier: Ambeed
Danger

A324403-1G A324403-25G A324403-5G
77601-452EA 90.2 USD
77601-452 77879-675 77879-676
Phenylselenol ≥98%
Phenylselenol

Precaución: for Research Use Only

Formula: C₆H₆Se
MDL Number: MFCD00000001
CAS Number: 645-96-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR