(6R,12aR)-2-Amino-6-(benzo[d][1,3]dioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione 95+%

Supplier: Ambeed
Warning

Synonyms: Amino Tadalafil

A324267-100MG A324267-10G A324267-10MG A324267-1G A324267-5G A324267-5MG
77612-870EA 20.6 USD
77612-870 77672-656 77672-658 77672-660 77672-662 77672-664
(6R,12aR)-2-Amino-6-(benzo[d][1,3]dioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione 95+%
(6R,12aR)-2-Amino-6-(benzo[d][1,3]dioxol-5-yl)-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione

Precaución: for Research Use Only

Formula: C₂₁H₁₈N₄O₄
MW: 390.39 g/mol
MDL Number: MFCD22690876
CAS Number: 385769-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR