2,4,5-Trifluorophenylboronic acid ≥98%

Supplier: Ambeed
Warning

A323841-1G A323841-5G A323841-25G
77593-922EA 13.33 USD
77593-922 77616-424 77748-636
2,4,5-Trifluorophenylboronic acid ≥98%
2,4,5-Trifluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₃O₂
MDL Number: MFCD01863165
CAS Number: 247564-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR