4-Bromo-1H-imidazole-2-carboxamide ≥95%
Supplier: Ambeed
|
Warning
|
A317587-250MG
A317587-1G
77612-836EA
226.6
USD
77612-836
77890-634
4-Bromo-1H-imidazole-2-carboxamide ≥95%
4-Bromo-1H-imidazole-2-carboxamide
Precaución: for Research Use Only
Formula:
C₄H₄BrN₃O MW: 190.00 g/mol |
CAS Number:
1823952-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...