6-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥96%, Ambeed.Inc

Supplier: Ambeed
Warning

A317387-5G A317387-1G A317387-250MG
77683-943EA 414 USD
77683-943 77892-654 77892-655
6-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥96%, Ambeed.Inc
6-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: For research use only.

Formula: C₉H₁₁Cl₂N
MDL Number: MFCD08437638
CAS Number: 33537-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR