3-(9-((4-((2,5-Dioxopyrrolidin-1-yl)oxy)-4-oxobutyl)(tosyl)carbamoyl)acridin-10-ium-10-yl)propane-1-sulfonate ≥97%

Supplier: Ambeed
Warning

Synonyms: NSP-SA-NHS

A317382-10MG A317382-1MG A317382-5MG
77616-390EA 175.1 USD
77616-390 77616-392 77616-394
3-(9-((4-((2,5-Dioxopyrrolidin-1-yl)oxy)-4-oxobutyl)(tosyl)carbamoyl)acridin-10-ium-10-yl)propane-1-sulfonate ≥97%
3-(9-((4-((2,5-Dioxopyrrolidin-1-yl)oxy)-4-oxobutyl)(tosyl)carbamoyl)acridin-10-ium-10-yl)propane-1-sulfonate

Precaución: for Research Use Only

Formula: C₃₂H₃₁N₃O₁₀S₂
MW: 681.73 g/mol
MDL Number: MFCD22200709
CAS Number: 199293-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR