Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III) ≥98%

Supplier: Ambeed
Warning

Synonyms: [(ppy)2IrCl]2, Dichlorotetrakis[2-(2-pyridinyl)phenyl]diiridium(III), Dichlorotetrakis(2-(2-pyridinyl)phenyl)diiridium(III)

A323115-250MG A323115-100MG A323115-1G
77571-912EA 55.62 USD
77571-912 77685-925 77685-926
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III) ≥98%
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

Precaución: for Research Use Only

Formula: C₄₄H₃₂Cl₂Ir₂N₄
MW: 1072.09 g/mol
MDL Number: MFCD07784515
CAS Number: 92220-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR