MS37452 ≥95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(2,3-Dimethoxybenzoyl)piperazin-1-yl)-2-(m-tolyloxy)ethanone

A317017-100MG A317017-50MG A317017-250MG
77748-617EA 101 USD
77748-617 77748-619 77748-618
MS37452 ≥95%
MS37452

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₂O₅
MW: 398.45 g/mol
MDL Number: MFCD03134667
CAS Number: 423748-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR