24-Crown-8-ether ≥98%

Supplier: Ambeed
Warning

Synonyms: 1,4,7,10,13,16,19,22-Octaoxacyclotetracosane

A322534-250MG A322534-1G A322534-50MG A322534-100MG
77746-626EA 87 USD
77746-626 77746-625 77746-627 77746-624
24-Crown-8-ether ≥98%
24-Crown-8-ether

Precaución: for Research Use Only

Formula: C₁₆H₃₂O₈
MW: 352.42 g/mol
MDL Number: MFCD00054528
CAS Number: 33089-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR