4-(Fluoromethyl)piperidin-4-ol hydrochloride ≥98%

Supplier: Ambeed
Warning

A322493-100MG A322493-250MG
77685-923EA 143 USD
77685-923 77685-924
4-(Fluoromethyl)piperidin-4-ol hydrochloride ≥98%
4-(Fluoromethyl)piperidin-4-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClFNO
MW: 169.62 g/mol
MDL Number: MFCD20688185
CAS Number: 1253929-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR