5-Fluoro-2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98%

Supplier: Ambeed
Warning

A316295-1G A316295-250MG A316295-5G
77612-828EA 459.38 USD
77612-828 77612-830 77612-832
5-Fluoro-2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98%
5-Fluoro-2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
MDL Number: MFCD18712646
CAS Number: 1788062-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR