2,6-Dimethyl-3-nitroaniline ≥97%

Supplier: Ambeed
Warning

A322248-1G A322248-250MG
77885-856EA 36 USD
77885-856 77885-857
2,6-Dimethyl-3-nitroaniline ≥97%
2,6-Dimethyl-3-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00156348
CAS Number: 67083-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR