1,3,5,7-Tetraphenyladamantane ≥97%

Supplier: Ambeed
Warning

A321985-5G A321985-25G
77593-914EA 138.38 USD
77593-914 77746-622
1,3,5,7-Tetraphenyladamantane ≥97%
1,3,5,7-Tetraphenyladamantane

Precaución: for Research Use Only

Formula: C₃₄H₃₂
MW: 440.62 g/mol
MDL Number: MFCD03104272
CAS Number: 16004-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR