Boc-Cys(stbu)-OH ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)-3-(tert-butyldisulphanyl)propanoic acid
A321915-1G
A321915-100MG
77626-594EA
72.1
USD
77626-594
77878-016
Boc-Cys(stbu)-OH ≥99%
Boc-Cys(stbu)-OH
Precaución: for Research Use Only
Formula:
C₁₂H₂₃NO₄S₂ |
MDL Number:
MFCD00084687 CAS Number: 30044-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...