5-Fluoro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A321764-100MG A321764-250MG
77872-454EA 27 USD
77872-454 77872-455
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid ≥98%
5-Fluoro-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅FN₂O₂
MDL Number: MFCD11518911
CAS Number: 920978-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR