Tris(2,4-di-tert-butylphenyl) phosphate ≥95%

Supplier: Ambeed
Warning

A315896-100MG A315896-1G A315896-250MG
77612-822EA 136.99 USD
77612-822 77683-939 77683-940
Tris(2,4-di-tert-butylphenyl) phosphate ≥95%
Tris(2,4-di-tert-butylphenyl) phosphate

Precaución: For research use only.

Formula: C₄₂H₆₃O₄P
MW: 662.92 g/mol
MDL Number: MFCD29918884
CAS Number: 95906-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR