Boc-L-Asp(OAll)-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: Boc-Asp(OAll)-OH, (S)-4-(Allyloxy)-2-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid

A321220-1G A321220-5G A321220-25G
77612-856EA 22.13 USD
77612-856 77612-858 77685-919
Boc-L-Asp(OAll)-OH ≥98%
Boc-L-Asp(OAll)-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₆
MDL Number: MFCD00273398
CAS Number: 132286-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR