tert-Butyl-1-pyrazolidinecarboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl pyrazolidine-1-carboxylate, Pyrazolidine, N1-BOC protected

A315442-100MG A315442-1G
77879-644EA 149 USD
77879-644 77879-645
tert-Butyl-1-pyrazolidinecarboxylate ≥95%
tert-Butyl-1-pyrazolidinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₂
MDL Number: MFCD09817426
CAS Number: 57699-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR