Ethyl-2-piperidinecarboxylate ≥98%

Supplier: Ambeed
Danger

A315397-1G A315397-25G
77746-603EA 6 USD
77746-603 77746-604
Ethyl-2-piperidinecarboxylate ≥98%
Ethyl 2-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD00005980
CAS Number: 15862-72-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR