(2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-methoxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥95%

Supplier: Ambeed
Warning

Synonyms: Desoxyrhaponticin

A315012-1MG A315012-5MG A315012-10MG
77616-382EA 43.26 USD
77616-382 77616-384 77616-380
(2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-methoxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol ≥95%
(2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-methoxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₂₁H₂₄O₈
MW: 404.41 g/mol
MDL Number: MFCD00063444
CAS Number: 30197-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR