Cephalexin monohydrate ≥98%

Supplier: Ambeed
Warning

A320894-100G A320894-25G
77601-446EA 147.6 USD
77601-446 77626-590
Cephalexin monohydrate ≥98%
Cephalexin monohydrate

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₄S
MDL Number: MFCD00167148
CAS Number: 23325-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR