Apatinib ≥98%

Supplier: Ambeed
Warning

Synonyms: Apatinib, tyrosine kinase inhibitor, N-(4-(1-Cyanocyclopentyl)phenyl)-2-((pyridin-4-ylmethyl)amino)nicotinamide, N-(4-(1-cyanocyclopentyl)phenyl)-2-((4-methylpyridin-3-yl)amino)nicotinamide, YN968D1

A314938-10MG A314938-1MG A314938-50MG A314938-5MG
77612-814EA 78.28 USD
77612-814 77612-816 77612-818 77612-820
Apatinib ≥98%
Apatinib

Precaución: for Research Use Only

Formula: C₂₄H₂₃N₅O
MW: 397.47 g/mol
MDL Number: MFCD22420814
CAS Number: 811803-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR