rel-(3S,4R)-1-(tert-Butoxycarbonyl)-4-(4-(trifluoromethyl)phenyl)pyrrolidine-3-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A314917-100MG A314917-1G
77612-812EA 62.83 USD
77612-812 77672-614
rel-(3S,4R)-1-(tert-Butoxycarbonyl)-4-(4-(trifluoromethyl)phenyl)pyrrolidine-3-carboxylic acid ≥95%
rel-(3S,4R)-1-(tert-Butoxycarbonyl)-4-(4-(trifluoromethyl)phenyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₀F₃NO₄
MW: 359.34 g/mol
MDL Number: MFCD02089478
CAS Number: 955137-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR