1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥98%

Supplier: Ambeed
Warning

Synonyms: DiR iodide, 1,1-Dioctadecyl-3,3,3,3-tetramethylindotricarbocyanine iodide, 2-(7-(3,3-Dimethyl-1-octadecylindolin-2-ylidene)hepta-1,3,5-trien-1-yl)-3,3-dimethyl-1-octadecyl-3H-indol-1-ium iodide, DiIC18(7)

A320307-50MG A320307-10MG A320307-100MG A320307-250MG
77616-414EA 141.11 USD
77616-414 77746-618 77746-617 77746-619
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥98%
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR)

Precaución: for Research Use Only

Formula: C₆₃H₁₀₁N₂I
MW: 1013.39 g/mol
MDL Number: MFCD03427788
CAS Number: 100068-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR