5-Amino-3,4-dimethyl-N-(4-((trifluoromethyl)sulfonyl)benzyl)thieno[2,3-c]pyridazine-6-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: VU0467154

A320248-10MG A320248-1MG A320248-5MG
77598-418EA 134.28 USD
77598-418 77598-420 77598-422
5-Amino-3,4-dimethyl-N-(4-((trifluoromethyl)sulfonyl)benzyl)thieno[2,3-c]pyridazine-6-carboxamide ≥98%
5-Amino-3,4-dimethyl-N-(4-((trifluoromethyl)sulfonyl)benzyl)thieno[2,3-c]pyridazine-6-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₅F₃N₄O₃S₂
MW: 444.45 g/mol
MDL Number: MFCD31615456
CAS Number: 1451993-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR